Learn More
Medchemexpress LLC Polydatin (Standard) | 27208-80-6 | 98.4% | 390.38 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN0120AR5MG
Encompass_Preferred
Polydatin (Standard) is the analytical standard of Polydatin, intended for research and analytical applications. Extracted from the roots of Reynoutria japonica, a widely used traditional Chinese remedy, it possesses anti-inflammatory activity in several experimental models and inhibits G6PD, inducing oxidative and ER stresses.
  • Analytical standard for Polydatin
  • Intended for research and analytical applications
  • Extracted from the roots of Reynoutria japonica
  • Possesses anti-inflammatory activity
  • Inhibits G6PD, inducing oxidative and ER stresses
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-30284