Alfa Aesar™ Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 139,000

Manufacturer: Alfa Aesar™ 44644A6

Quantity 4kg

 View more versions of this product

Catalog No. AA44644A6

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 9016-00-6
    Molecular Formula (CH3)3SiO(Si(CH3)2O)nSi(CH3)3
    MDL Number MFCD00148360
    Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 139,000

    Specifications

    Physical Form Viscous Liquid
    Melting Point <−40°C
    Density 0.977g/mL
    Molecular Formula (CH3)3SiO(Si(CH3)2O)nSi(CH3)3
    CAS 9016-00-6
    Odor Odorless
    Quantity 4kg
    Flash Point 321°C (610°F)
    MDL Number MFCD00148360
    Refractive Index 1.4035
    Boiling Point >205°C
    Color Colorless
    RTECS Number TQ2690000
    Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 139,000

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit