Alfa Aesar™ Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 2000

Manufacturer: Alfa Aesar™ 42499K2

Quantity 1L

 View more versions of this product

Catalog No. AA42499K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 9016-00-6
    MDL Number MFCD00148360
    Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 2,000

    Specifications

    Melting Point <−40°C
    Density 0.966g/mL
    Physical Form Liquid
    MDL Number MFCD00148360
    Odor Odorless
    Refractive Index 1.4000
    Boiling Point >205°C
    Color Colorless
    Flash Point 232°C (450°F)
    CAS 9016-00-6
    Quantity 1L
    RTECS Number TQ2690000
    Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 2,000

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit