Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 237, Thermo Scientific Chemicals

Catalog Number AA42496K2
Click to view available options
Quantity:
50 mL
250 mL
1 L
9016-00-6
(C2H6OSi)n
74.15
MFCD00084411
SEUDSDUUJXTXSV-UHFFFAOYSA-N
Hexamethyldisiloxane polymers
dimethylsilanone
C[Si](C)(-*)O-*
9016-00-6
(C2H6OSi)n
74.15
MFCD00084411
SEUDSDUUJXTXSV-UHFFFAOYSA-N
Hexamethyldisiloxane polymers
dimethylsilanone
C[Si](C)(-*)O-*

Polydimethylsiloxane, trimethylsiloxy is used in water-repellent, protective creams & lotions. It is also used as surfactant, is used in coating auxiliary agents. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Physical Form Liquid
Boiling Point 152°C to 153°C
Melting Point -86°C
UN Number UN1993
Density 0.820 g/mL
Solubility Information Soluble in aromatic hydrocarbon,partially soluble in aliphatic and insoluble in water.
Quantity 1 L
Flash Point 39°C (102°F)
Odor Odorless
Refractive Index 1.3825
Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 237
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECSNumber TQ2690000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit