Alfa Aesar™ Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 410

Manufacturer: Alfa Aesar™ 43669AD

Quantity 50mL

 View more versions of this product

Catalog No. AA43669AD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 9016-00-6
    Molecular Formula (CH3)3SiO(Si(CH3)2O))nSi(CH3)3
    MDL Number MFCD00148360
    Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 410

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 9016-00-6
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Refractive Index 1.3900
    Quantity 50mL
    Molecular Formula (CH3)3SiO(Si(CH3)2O))nSi(CH3)3
    MDL Number MFCD00148360
    Odor Odorless
    Boiling Point 85° to 105°C (2° to 4mmHg)
    Flash Point 70°C (158°F)
    RTECS Number TQ2690000
    Density 0.88g/mL
    Physical Form Liquid
    Color Colorless
    Melting Point −40°C
    Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 410

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit