Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Polyester Adhesive Warning Sign: HAZARDOUS MATERIALS MAY BE USED IN THIS AREA SEE MSDS

Improve workplace safety with proper signage and markings. This Brady™ self-adhesive mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.

$14.13 - $33.96

Specifications

Color Black on Yellow
Message HAZARDOUS MATERIALS MAY BE USED IN THIS AREA SEE MSDS
Quantity 1 Ea.
Shape Rectangular
Thickness 0.01 in.
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-105-560
View Documents
Brady™
126002
10 x 7 in.
Each for $14.13
Add to cart
 
19-105-563
View Documents
Brady™
126005
14 x 10 in.
Each for $33.96
Add to cart
 
Description

Description

  • Polyester material is abrasion-resistant and chemical-resistant
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
Specifications

Specifications

Black on Yellow
1 Ea.
0.01 in.
Polyester
HAZARDOUS MATERIALS MAY BE USED IN THIS AREA SEE MSDS
Rectangular
Cold temperature permanent acrylic
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit