Learn More
Brady™ Polyester Adhesive Warning Sign: PESTICIDE STORAGE AREA ALL UNAUTHORIZED PERSONS KEEP OUT FIRE MAY CAUSE TOXIC FUMES
Improve workplace safety with proper signage and markings. This Brady™ self-adhesive mounted sign is a durable, clear and effective tool to raise awareness to chemical, biohazard and hazardous material warnings.
$18.64 - $44.40
Specifications
Color | Red on White |
---|---|
Shape | Rectangular |
Displayed Text | PESTICIDE STORAGE AREA ALL UNAUTHORIZED PERSONS KEEP OUT FIRE MAY CAUSE TOXIC FUMES |
Temperature Range (English) | -40°F to 230°F |
Finish | Laminated |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-107-465
|
Brady™
84430 |
10 x 7 in. |
Each for $18.64
|
|
|||||
19-107-466
|
Brady™
84431 |
14 x 10 in. |
Each for $44.40
|
|
|||||
Description
- Polyester material is abrasion-resistant and chemical-resistant
- Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- For indoor and outdoor use
- Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
Specifications
Red on White | |
PESTICIDE STORAGE AREA ALL UNAUTHORIZED PERSONS KEEP OUT FIRE MAY CAUSE TOXIC FUMES | |
Laminated | |
1 Ea. | |
Self-adhesive | |
-40°F |
Rectangular | |
-40°F to 230°F | |
Polyester | |
Cold-temperature Permanent Acrylic | |
230°F | |
0.01 in. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.