Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Polystyrene ANSI Sign: CONFINED SPACE. ENTER BY PERMIT ONLY. PREPARE FOR ENTRY. TEST ATMOSPHERE. PREPARE PERSONAL PROTECTION DEVICES.
Improve workplace safety with non-adhesive mounted signage. The Brady™ Confined Space ANSI Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.
$18.11 - $44.40
Catalog Number | Mfr. No. | Message | Dimensions (L x W) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Message | Dimensions (L x W) | Price | Quantity & Availability | ||||
19-107-820
|
Brady™
143672 |
CONFINED SPACE. ENTER BY PERMIT ONLY. PREPARE FOR ENTRY. TEST ATMOSPHERE. PREPARE PERSONAL PROTECTION DEVICES. | 14 x 10 in. |
Each for $28.35
|
|
19-107-821
|
Brady™
143673 |
CONFINED SPACE. ENTER BY PERMIT ONLY. PREPARE FOR ENTRY. TEST ATMOSPHERE. PREPARE PERSONAL PROTECTION DEVICES. | 10 x 7 in. |
Each for $18.11
|
|
19-112-867
|
Brady™
144764 |
TRIPPING HAZARD. FALLS MAY RESULT IN INJURY. | 10 x 7 in. |
Each for $18.64
|
|
19-112-868
|
Brady™
144765 |
TRIPPING HAZARD. FALLS MAY RESULT IN INJURY. | 14 x 10 in. |
Each for $44.40
|
|
Description
- Polystyrene construction is abrasion-resistant and chemical-resistant
- Corner holes provide easy mounting solutions in a variety of environments
Specifications
Rectangular | |
1 Ea. | |
-40°F |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title