Learn More
Brady™ Polyester Danger Sign: FUEL
Improve workplace safety with proper signage. The Brady™ Danger Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.
$14.13 - $33.96
Specifications
Adhesive Type Cold-temperature Permanent Acrylic
Color Black/Red on White
Description Polyester with polyester overlaminate
Message FUEL
Quantity 1 Ea.
View More Specs
Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-107-736
View Documents Promotion Details
Brady™
95002
10 x 7 in.
Each for $14.13
Add to cart
19-107-737
View Documents Promotion Details
Brady™
95003
14 x 10 in.
Each for $33.96
Add to cart
Description
Description
  • Polyester material is abrasion-resistant and chemical-resistant
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Bold and bright, providing high visibility to critical messages
Specifications
Specifications
Cold-temperature Permanent Acrylic
Polyester with polyester overlaminate
1 Ea.
0.01 in.
Black/Red on White
FUEL
Rectangular
Polyester