Learn More
Brady™ Polyester Danger Sign: LIQUID NITROGEN
Improve workplace safety with proper signage. The Brady™ Danger Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.
$11.75 - $21.99
Specifications
Adhesive Type Cold-temperature Permanent Acrylic
Color Black/Red on White
Description Polyester with polyester overlaminate
Message LIQUID NITROGEN
Quantity 1 Ea.
View More Specs
Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-105-821
View Documents
Brady™
126320
10 x 7 in.
Each for $11.75
Add to cart
19-105-824
View Documents
Brady™
126323
14 x 10 in.
Each for $21.99
Add to cart
Description
Description
  • Polyester material is abrasion-resistant and chemical-resistant
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Bold and bright, providing high visibility to critical messages
Specifications
Specifications
Cold-temperature Permanent Acrylic
Polyester with polyester overlaminate
1 Ea.
0.01 in.
Black/Red on White
LIQUID NITROGEN
Rectangular
Polyester