Learn More
Brady™ Polyester Encased Paper Tag: CAUTION DO NOT CLOSE THIS VALVE SIGNED BY: DATE:
Identify facility equipment to your exact specifications. The Brady™ Polyester Encased Paper Tag: CAUTION are durable two-sided tags with a write-on area that accepts most marking pens.
Manufacturer:  Brady™ 86427
Catalog No. 19-113-396
Add to cart
Description
Description
  • Metal grommet prevents tearing from the point of attachment
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use
  • Tags are abrasion and chemical resistant
Specifications
Specifications
Black on yellow
Matte
(10) nylon fasteners
10/Pack
-40°F to 176°F
DO NOT CLOSE THIS VALVE SIGNED BY: DATE:
3 in. x 5.75 in.
General equipment
Laminated
Rectangle
Polyester encased paper
Header: CAUTION