Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Polyester Encased Paper Tag: CAUTION OUT OF ORDER SIGNED BY: DATE:

Identify facility equipment to your exact specifications. The Brady™ Polyester Encased Paper Tag: CAUTION are durable two-sided tags with a write-on area that accepts most marking pens.

$36.31 - $38.73

Specifications

Color Black on yellow
Material Polyester encased paper
Finish Matte
Includes (10) nylon fasteners
Label Type Laminated
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-113-305
View Documents
Brady™
86424
3 in. x 5.75 in.
Pack of 10 for $36.31
Add to cart
 
19-113-308
View Documents
Brady™
86485
4 in. x 7 in.
Pack of 10 for $38.73
Add to cart
 
Description

Description

  • Metal grommet prevents tearing from the point of attachment
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use
  • Tags are abrasion and chemical resistant
Specifications

Specifications

Black on yellow
Matte
Laminated
Rectangle
Polyester encased paper
(10) nylon fasteners
10/Pack
-40°F to 176°F
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit