Learn More
Brady™ Polyester Encased Paper Tag: CONFINED SPACE ENTRY PERMIT
Identify facility equipment to your exact specifications. Brady™ Polyester Encased Paper Tags CONFINED SPACE ENTRY PERMIT are durable two-sided tags with a write-on area that accepts most marking pens.
Manufacturer:  Brady™ 49234
Catalog No. 19-113-454
Add to cart
Description
Description
  • Metal grommet prevents tearing from the point of attachment
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use
  • Tags are abrasion and chemical resistant
Specifications
Specifications
Black/red on white
Matte
(25) nylon fasteners
25/Pack
-40°F to 176°F
DATE: TIME:
3 in. x 5.75 in.
General equipment
Laminated
Rectangle
Polyester encased paper
Header: CONFINED SPACE ENTRY PERMIT