Learn More
Brady™ Polyester Encased Paper Tag: DANGER/PELIGRO DO NOT START THIS MACHINE INO PONGA EN MARCHA ESTA MAQUINA! SIGNED BY FIRMADO POR: DATE FECHA:
Identify facility equipment to your exact specifications. The Brady™ Polyester Encased Paper Tag: CAUTION are durable two-sided tags with a write-on area that accepts most marking pens.
Manufacturer:  Brady™ 86467
Catalog No. 19-113-399
Add to cart
Description
Description
  • Metal grommet prevents tearing from the point of attachment
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use
  • Tags are abrasion and chemical resistant
Specifications
Specifications
Black/red on white
Matte
(10) nylon fasteners
10/Pack
-40°F to 176°F
DO NOT START THIS MACHINE INO PONGA EN MARCHA ESTA MAQUINA! SIGNED BY FIRMADO POR: DATE FECHA:
4 in. x 7 in.
General equipment
Laminated
Rectangle
Polyester encased paper
Header: DANGER/PELIGRO