Learn More
Brady™ Polyester Encased Paper Tag: QUALITY CONTROL
Identify facility equipment to your exact specifications. Brady™ Polyester Encased Paper Tags QUALITY CONTROL are durable two-sided tags with a write-on area that accepts most marking pens.
Manufacturer:  Brady™ 86448
Catalog No. 19-113-339
Add to cart
Description
Description
  • Metal grommet prevents tearing from the point of attachment
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use
  • Tags are abrasion and chemical resistant
Specifications
Specifications
Red on white
Matte
Laminated
Rectangle
Polyester encased paper
3 in. x 5.75 in.
(10) nylon fasteners
10/Pack
-40°F to 176°F
Header: QUALITY CONTROL