Learn More
Brady™ Polyester Isolation Blind Tag: ISOLATION BLIND
Identify facility equipment to your exact specifications. Brady™ Polyester Isolation Blind Tags are durable one-sided tags that are an economical and convenient way to ID equipment.
Manufacturer:  Brady™ 132454
Catalog No. 19-113-438
Add to cart
Description
Description
  • Polyester tags provide identification solutions for almost any environment
  • Metal grommet prevents tearing from the point of attachment
  • Tags are appropriate for indoor or outdoor use
  • Tags are abrasion and chemical resistant
Specifications
Specifications
OSHA 1910.145
20 in. x 2 in.
(25) nylon fasteners
25/Pack
-40°F to 212°F
ISOLATION BLIND
Black on red
General equipment
Not laminated
Rectangle
Polyester