Learn More
PIP™ Polyester-Lined Hot Mill Glove
PIP™ Polyester-Lined Hot Mill Glove is used in Glass/Plastics Manufacturing, Foundry Operations, Steel Mills, Automotive
Manufacturer:  PIP™ B03SI
Catalog No. 19-050-203
Add to cart
Description
Description
  • Extra heavy weight
  • Two-layer cotton/polyester construction offers comfort, insulation and superior heat protection
  • 100% cotton outer shell|
  • Clute cut provides a roomy fit
  • Straight thumb increases comfort in closed-fisted work
  • Nap-out finish for heat resistance
  • Knuckle strap for extra protection
  • Band Top cuff helps protect wrist area and allows for easy on and off
Specifications
Specifications
Band Top Cuff
Large
Natural
Cotton, Polyester
30 oz.
None