Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Polyester Permanent Rubber Adhesive BradyGlo™ Exit & Entrance Sign: NOT AN EXIT

Improve workplace safety with proper signage and markings. This Brady™ self-adhesive mounted sign is a durable, clear and effective tool to clearly mark exits and entrances.

$10.25 - $71.50

Specifications

Color Black on glow
Material Polyester
Message NOT AN EXIT
Min. Temperature (English) -40°F
Mounting Type Self-adhesive
View More Specs

Products 2
Catalog Number Mfr. No. Style Price Quantity & Availability  
Catalog Number Mfr. No. Style Price Quantity & Availability  
19-109-800 View Documents Brady™
81712
Pictogram #1 Each for $10.25
Add to cart
 
19-109-801 View Documents Brady™
81713
Pictogram #2 Pack of 10 for $71.50
Add to cart
 
Description

Description

  • Phosphorescent polyester material is abrasion-resistant and chemical-resistant
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • For indoor use
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
Specifications

Specifications

Black on glow
NOT AN EXIT
Self-adhesive
3.5 in.
10 in. x 3.5 in. x 0.0094 in.
Polyester
-40°F
10 in.
Rectangle
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit