Learn More
Brady™ Polyester Sign: DANGER: DO NOT ENTER WITHOUT WEARING EAR PROTECTION. SEVERE HEARING DAMAGE WILL RESULT.
Improve workplace safety with proper signage and markings. This Brady™ polyester sign is a durable, clear and effective tool to raise awareness to workplace hazards.
Manufacturer:  Brady™ 144307
 View more versions of this product
Catalog No. 19-111-758
Add to cart
Description
Description
  • Polyester material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Specifications
Red, Blue, Black on White
Rectangle
DANGER: DO NOT ENTER WITHOUT WEARING EAR PROTECTION. SEVERE HEARING DAMAGE WILL RESULT.
Polyester
14 in. x 10 in. x 0.01 in.