Learn More
Brady™ Polyester Sign: DO NOT HANDLE WITHOUT WEARING SAFETY SHIELD. ACCIDENT WILL RESULT IN SERIOUS INJURY OR DEATH.
Improve workplace safety with proper signage and markings. This Brady™ polyester sign is a durable, clear and effective tool to raise awareness to workplace hazards.
Manufacturer:  Brady™ 144210
 View more versions of this product
Catalog No. 19-111-687
Add to cart
Description
Description
  • Polyester material is abrasion-resistant and chemical-resistant
  • Safety signs are important facility visuals that are vital to communicating warnings and other safety information
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Bold and bright, providing high visibility to critical messages
  • For indoor and outdoor use
Specifications
Specifications
Red, Blue, Black on White
Rectangle
DO NOT HANDLE WITHOUT WEARING SAFETY SHIELD. ACCIDENT WILL RESULT IN SERIOUS INJURY OR DEATH.
Polyester
14 in. x 10 in. x 0.01 in.