Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Polyester Sign: DO NOT WATCH ARC. VISION DAMAGE MAY OCCUR.
Improve workplace safety with proper signage and markings. This Brady™ polyester sign is a durable, clear and effective tool to raise awareness to workplace hazards.
$29.50 - $30.00
Specifications
Material | Polyester |
---|---|
Shape | Rectangular |
Displayed Text | DO NOT WATCH ARC. VISION DAMAGE MAY OCCUR. |
Color | Yellow on White |
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Price | Quantity & Availability | |||||
19-112-556
|
Brady™
144243 |
7 x 10 in. |
Each for $29.50
|
|
|||||
19-112-559
|
Brady™
144246 |
10 x 14 in. |
Each for $30.00
|
|
|||||
Description
- Polyester material is abrasion-resistant and chemical-resistant
- Safety signs are important facility visuals that are vital to communicating warnings and other safety information
- Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
- Bold and bright, providing high visibility to critical messages
- For indoor and outdoor use
Specifications
Polyester | |
Rectangular | |
DO NOT WATCH ARC. VISION DAMAGE MAY OCCUR. | |
Yellow on White |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title