Learn More
Brady™ Polyester Tag: CALIBRATED
Identify facility equipment to your exact specifications. Brady™ Polyester Tags CALIBRATED are durable one-sided tags with a write-on area that accepts most marking pens.
Manufacturer:  Brady™ 76158
Catalog No. 19-113-337
Add to cart
Description
Description
  • Polyester tags provide identification solutions for almost any environment
  • Metal grommet prevents tearing from the point of attachment
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use
  • Tags are abrasion and chemical resistant
Specifications
Specifications
OSHA 1910.145
5.75 in. x 3 in.
Not laminated
Rectangle
Polyester
Header: CALIBRATED
Black on white
(25) nylon fasteners
25/Pack
-40°F to 212°F
REMARKS: SIGNED BY: DATE: