Learn More
Brady™ Polyester Tag: CAUTION UNSAFE DO NOT USE SIGNED BY: DATE:
Identify facility equipment to your exact specifications. Brady™ Polyester Tags are durable two-sided tags with a write-on area that accepts most marking pens.
Manufacturer:  Brady™ 48141
Catalog No. 19-113-360
Add to cart
Description
Description
  • Polyester tags provide identification solutions for almost any environment
  • Metal grommet prevents tearing from the point of attachment
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use
  • Tags are abrasion and chemical resistant
Specifications
Specifications
OSHA 1910.145
3 in. x 5.75 in.
(10) nylon fasteners
10/Pack
-40°F to 212°F
UNSAFE DO NOT USE SIGNED BY: DATE:
Black on yellow
General equipment
Not laminated
Rectangle
Polyester
Header: CAUTION