Learn More
Brady™ Polyester Tag: DANGER DO NOT OPERATE ELECTRICAL DEPARTMENT SIGNED BY: DATE:
Identify facility equipment to your exact specifications. Brady™ Polyester Tags are durable two-sided tags with a write-on area that accepts most marking pens.
Manufacturer:  Brady™ 76178
Catalog No. 19-113-387
Add to cart
Description
Description
  • Polyester tags provide identification solutions for almost any environment
  • Metal grommet prevents tearing from the point of attachment
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use
  • Tags are abrasion and chemical resistant
Specifications
Specifications
Header: DANGER
Rectangle
Polyester
OSHA 1910.145
3 in. x 5.75 in.
(25) nylon fasteners
25/Pack
-40°F to 212°F
DO NOT OPERATE ELECTRICAL DEPARTMENT SIGNED BY: DATE:
Black/red on white
General equipment
Not laminated