Learn More
Brady™ Polyester Tag: EMERGENCY SHOWER & EYEWASH TEST RECORD
Identify facility equipment to your exact specifications. Brady™ Polyester Tags EMERGENCY SHOWER & EYEWASH TEST RECORD are durable two-sided tags with a write-on area that accepts most marking pens.
Manufacturer:  Brady™ 76195
Catalog No. 19-113-136
Add to cart
Description
Description
  • Polyester tags provide identification solutions for almost any environment
  • Metal grommet prevents tearing from the point of attachment
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use
  • Tags are abrasion and chemical resistant
Specifications
Specifications
OSHA 1910.145
3 in. x 5.75 in.
(25) nylon fasteners
Rectangle
Polyester
Header: EMERGENCY SHOWER & EYEWASH TEST RECORD
Black/green on white
Emergency shower and eye wash stations
25/Pack
-40°F to 212°F
INSPECT THIS UNIT CAREFULLY BEFORE SIGNING INSPECTION RECORD DATE: BY: DATE: BY: