Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Polyester Tag: SCAFFOLD INSPECTION DATE: BY: DATE: BY:

Identify facility equipment to your exact specifications. Brady™ Polyester Tags SCAFFOLD INSPECTION are durable two-sided tags with a write-on area that accepts most marking pens.

Manufacturer:  Brady™ 86566

Catalog No. 19-113-141


Add to cart

Description

Description

  • Polyester tags provide identification solutions for almost any environment
  • Metal grommet prevents tearing from the point of attachment
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use
  • Tags are abrasion and chemical resistant
Specifications

Specifications

OSHA 1910.145
3 in. x 5.75 in.
(10) nylon fasteners
Rectangle
Polyester
Header: SCAFFOLD INSPECTION
Green on white
Scaffolding
10/Pack
-40°F to 212°F
DATE: BY: DATE: BY:
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit