Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Polyester Warning Sign: CONTAINS CADMIUM POISONOUS FUMES MAY BE FORMED ON HEATING

Improve workplace safety with proper signage. The Brady™ Warning Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.

$11.25 - $25.75

Specifications

Adhesive Type Cold-temperature Permanent Acrylic
Color Black on Orange
Description Polyester with polyester overlaminate
Message CONTAINS CADMIUM POISONOUS FUMES MAY BE FORMED ON HEATING
Quantity 1 Ea.
View More Specs

Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-105-985
View Documents
Brady™
126518
10 x 7 in. Each for $11.25
Add to cart
 
19-105-988
View Documents
Brady™
126521
14 x 10 in. Each for $25.75
Add to cart
 
Description

Description

  • Polyester material is abrasion-resistant and chemical-resistant
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Bold and bright, providing high visibility to critical messages
Specifications

Specifications

Cold-temperature Permanent Acrylic
Polyester with polyester overlaminate
1 Ea.
0.01 in.
Black on Orange
CONTAINS CADMIUM POISONOUS FUMES MAY BE FORMED ON HEATING
Rectangular
Polyester
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit