Learn More
Brady™ Polyester Warning Sign: DETECTABLE AMOUNTS OF CHEMICALS...THIS FACILITY.
Improve workplace safety with proper signage. The Brady™ Warning Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.
$11.25 - $25.75
Specifications
Adhesive Type Cold-temperature Permanent Acrylic
Color Black/Orange/Yellow on White
Description Polyester with polyester overlaminate
Message DETECTABLE AMOUNTS OF CHEMICALS KNOWN TO THE STATE OF CALIFORNIA TO CAUSE CANCER, BIRTH DEFECTS OR OTHER REPRODUCTIVE HARM MAY BE FOUND IN AND AROUND THIS FACILITY.
Quantity 1 Ea.
View More Specs
Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-106-455
View Documents
Brady™
144955
10 x 7 in.
Each for $11.25
Add to cart
19-106-456
View Documents
Brady™
144956
14 x 10 in.
Each for $25.75
Add to cart
Description
Description
  • Polyester material is abrasion-resistant and chemical-resistant
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Bold and bright, providing high visibility to critical messages
Specifications
Specifications
Cold-temperature Permanent Acrylic
Polyester with polyester overlaminate
1 Ea.
0.01 in.
Black/Orange/Yellow on White
DETECTABLE AMOUNTS OF CHEMICALS KNOWN TO THE STATE OF CALIFORNIA TO CAUSE CANCER, BIRTH DEFECTS OR OTHER REPRODUCTIVE HARM MAY BE FOUND IN AND AROUND THIS FACILITY.
Rectangular
Polyester