Learn More
Brady™ Polyester Warning Sign: PESTICIDE STORAGE AREA
Improve workplace safety with proper signage. The Brady™ Warning Sign is a durable, clear and effective tool to properly mark appropriate areas and to help prevent workplace injury.
$11.25 - $25.75
Specifications
Adhesive Type Cold-temperature Permanent Acrylic
Color Black on Orange
Description Polyester with polyester overlaminate
Message PESTICIDE STORAGE AREA
Quantity 1 Ea.
View More Specs
Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
19-106-056
View Documents
Brady™
126596
10 x 7 in.
Each for $11.25
Add to cart
19-106-059
View Documents
Brady™
126599
14 x 10 in.
Each for $25.75
Add to cart
Description
Description
  • Polyester material is abrasion-resistant and chemical-resistant
  • Printed on polymer plastic and overlaminated with a clear plastic coating to protect graphics against abrasion, sunlight, chemicals, and weather up to 8 years outdoors, and even longer indoors
  • Bold and bright, providing high visibility to critical messages
Specifications
Specifications
Cold-temperature Permanent Acrylic
Polyester with polyester overlaminate
1 Ea.
0.01 in.
Black on Orange
PESTICIDE STORAGE AREA
Rectangular
Polyester