Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Mopec Polyethylene Body Tray Available on GSA/VA Contract for Federal Government customers only.

Heavy-duty all white body tray with uncompromising construction for extra support. Mopec™ Polyethylene Body Tray that is specifically designed for mortuary and post mortem x-rays.

Manufacturer:  Mopec JC200

Catalog No. 22-444-173



Description

Description

  • Constructed of HDPE (High Density Polyethylene)
  • White in color

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Warranty and Services

1 Year Warranty

Specifications

Specifications

Holds up to 350lbs
Holds up to 158.76kg
23.12 in.
58.72cm
Transport and Handling
Body Trays
High Density Polyethylene
77.5 in.
196.85cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit