Alfa Aesar™ Polyethylene, chlorinated

Manufacturer: Alfa Aesar™ 44146A1

Quantity 1kg

 View more versions of this product

Catalog No. AA44146A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 64754-90-1
    MDL Number MFCD00084424
    Chemical Name or Material Polyethylene, Chlorinated

    Specifications

    TSCA TSCA
    CAS 64754-90-1
    Color Off-white
    RTECS Number KM6893750
    Physical Form Granular
    MDL Number MFCD00084424
    Quantity 1kg
    Chemical Name or Material Polyethylene, Chlorinated

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit