Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene glycol 10,000, Alfa Aesar

Click to view available options
Quantity:
250g
1000g
25322-68-3
H(OCH2CH2)nOH
25322-68-3
H(OCH2CH2)nOH
Density 1.2
Formula Weight ≈10000
Physical Form Powder
Quantity 1000g
Chemical Name or Material Polyethylene glycol 10, 000