Polyethylene glycol 10,000, Alfa Aesar

Manufacturer: Alfa Aesar B2195530

Quantity 250g

 View more versions of this product

Catalog No. AAB2195530

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25322-68-3
    Physical Form Powder
    Molecular Formula H(OCH2CH2)nOH
    Formula Weight ≈10,000
    Chemical Name or Material Polyethylene glycol 10, 000

    Specifications

    Formula Weight ≈10,000
    Quantity 250g
    Physical Form Powder
    Density 1.2
    Molecular Formula H(OCH2CH2)nOH
    CAS 25322-68-3
    Chemical Name or Material Polyethylene glycol 10, 000

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit