Polyethylene glycol 1000, Alfa Aesar

Manufacturer: Alfa Aesar B221340B

Quantity 1000g

 View more versions of this product

Catalog No. AAB221340B

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 25322-68-3
    Physical Form Powder/Low Melting Solid
    Molecular Formula H(OCH2CH2)nOH
    Chemical Name or Material Polyethylene glycol 1000

    Specifications

    Physical Form Powder/Low Melting Solid
    Quantity 1000g
    Density 1.101
    Molecular Formula H(OCH2CH2)nOH
    CAS 25322-68-3
    Chemical Name or Material Polyethylene glycol 1000

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit