Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol 1000, Waxy Soft Solid, BAKER™, J.T.Baker™

Click to view available options
Quantity:
500mL
Packaging:
Poly Bottle

Chemical Identifiers

25322-68-3

A grade of chemicals for laboratory use

CAS 25322-68-3
Formula Weight 950 to 1050 (Average)
Physical Form Waxy Soft Solid
Packaging Poly Bottle
Specific Gravity 1L = 1.15kg
Quantity 500mL
Chemical Name or Material Polyethylene Glycol 1000