Polyethylene Glycol 20,000, Flake, BAKER™, J.T.Baker™

A grade of chemicals for laboratory use

Manufacturer: J.T.Baker U20408

Packaging Poly Bottle
Quantity 2kg

 View more versions of this product

Catalog No. 02-003-640

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 25322-68-3
    Formula Weight 15,000 to 20,000 (Average)
    Chemical Name or Material Polyethylene Glycol 20,000

    Specifications

    Physical Form Flakes
    Formula Weight 15,000 to 20,000 (Average)
    Quantity 2kg
    CAS 25322-68-3
    Packaging Poly Bottle
    Chemical Name or Material Polyethylene Glycol 20,000

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit