Alfa Aesar™ Polyethylene glycol 4-tert-octylphenyl ether

Manufacturer: Alfa Aesar™ H327880F

Quantity 2500mL

 View more versions of this product

Catalog No. AAH327880F

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    MDL Number MFCD01779734
    Chemical Name or Material Polyethylene glycol 4-tert-octylphenyl ether

    Specifications

    MDL Number MFCD01779734
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H302
    Causes serious eye damage.
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P280d-P273-P305+P351+P338
    Wear protective clothing and eye protection.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Boiling Point >250°C
    Melting Point ≈5°C
    Quantity 2500mL
    Flash Point 180°C (356°F)
    Chemical Name or Material Polyethylene glycol 4-tert-octylphenyl ether

    GHS H Statement
    H318-H302
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P280d-P273-P305+P351+P338
    Wear protective clothing and eye protection.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit