Polyethylene glycol 6,000

Manufacturer: Alfa Aesar A1754130

Quantity 250g

 View more versions of this product

Catalog No. AAA1754130

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25322-68-3
    Molecular Formula H(OCH2CH2)nOH
    MDL Number MFCD01779614
    Merck Index 14,7568
    Chemical Name or Material Polyethylene Glycol 6000

    Specifications

    Quantity 250g
    TSCA TSCA
    MDL Number MFCD01779614
    Merck Index 14,7568
    RTECS Number TQ4100000
    Physical Form Powder
    Molecular Formula H(OCH2CH2)nOH
    CAS 25322-68-3
    Chemical Name or Material Polyethylene Glycol 6000

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit