Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene glycol 600, Alfa Aesar™

Click to view available options
Quantity:
250g
1000g
25322-68-3
H(OCH2CH2)nOH
MFCD00081839
25322-68-3
H(OCH2CH2)nOH
MFCD00081839
Density 1.12
Solubility Information Miscible with water,many organic solvents,aliphatic ketones,alcohols,chloroform,glycol ethers,esters,and aromatic hydrocarbons. Immiscible with ether and aliphatic hydrocarbons.
Refractive Index 1.468
Melting Point 17°C to 23°C
Quantity 1000g
Flash Point 252°C (485°F)
Chemical Name or Material Polyethylene glycol 600

P210-P233-P240-P241-P242-P243-P264b-P280-P302+P352-P304+P340-P305+P351+P338-P332+P313-P362-P374-P501c

H227-H303-H315-H320-H333-H335

EINECSNumber : 500-038-2

RTECSNumber : TQ3800000

TSCA : Yes

Recommended Storage : Ambient temperatures