Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol 6000, Flakes, Ultrapure, Affymetrix/USB™

Click to view available options
Quantity:
1kg
5kg
Packaging:
Plastic bottle
25322-68-3
MFCD01779614
PEG 6000
25322-68-3
MFCD01779614
PEG 6000
  • Differential precipitation of DNA
  • Enhances hybridization rate of nucleic acids
  • Improves efficiency of end-labeling with T4 Polynucleotide Kinase(1,2)
  • Used in ligation of blunt ended DNA
  • Precipitates bacteriophage &
Formula Weight 5400 to 6600
Physical Form Flakes
Packaging Plastic bottle
Melting Point 60°C to 63°C
Quantity 5kg
Chemical Name or Material Polyethylene Glycol 6000