Alfa Aesar™ Polyethylene glycol 8,000

Manufacturer: Alfa Aesar™ 4344336

Quantity 500g

 View more versions of this product

Catalog No. AA4344336

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25322-68-3
    Molecular Formula H(OCH2CH2)nOH
    MDL Number MFCD01779615
    Merck Index 14,7568
    Synonym PEG 8000
    Chemical Name or Material Polyethylene glycol 8,000

    Specifications

    Physical Form Powder
    Synonym PEG 8000
    CAS 25322-68-3
    Odor Odorless
    Color White
    pH 5 to 7
    Quantity 500g
    TSCA TSCA
    Molecular Formula H(OCH2CH2)nOH
    MDL Number MFCD01779615
    Merck Index 14,7568
    Melting Point 55° to 63°C
    RTECS Number TQ4105000
    Chemical Name or Material Polyethylene glycol 8,000

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit