Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Polyethylene Glycol 8000, NF, Spectrum™ Chemical
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Description
Spectrum™ Chemical Polyethylene Glycol 8000, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments. All Spectrum Chemical NF products, including all NF polyethylene glycol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
pH | 4.5 to 7.5 |
CAS Max % | 1 |
Quantity | 500 g |
Grade | NF |
Ignition Residue | 0.001 |
Packaging | Poly Bottle |
Viscosity | 470-900 cSt |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction