Alfa Aesar™ Polyethylene glycol di-p-toluenesulfonate, M.W. 3,400

Manufacturer: Alfa Aesar™ 4695906

Quantity 5g

 View more versions of this product

Catalog No. AA4695906

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula CH3C6H4SO2(OCH2CH2)nOSO2C6H4CH3
    Synonym PEG-di-p-tosylate
    Chemical Name or Material Polyethylene glycol di-p-toluenesulfonate, M.W. 3,400

    Specifications

    Physical Form Granular Solid
    Molecular Formula CH3C6H4SO2(OCH2CH2)nOSO2C6H4CH3
    Melting Point 43° to 47°C
    Quantity 5g
    Synonym PEG-di-p-tosylate
    Color White to off-white
    Chemical Name or Material Polyethylene glycol di-p-toluenesulfonate, M.W. 3,400

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit