Alfa Aesar™ Polyethylene glycol diacrylate, M.W. 8,000

Manufacturer: Alfa Aesar™ 4680103

Quantity 1g

 View more versions of this product

Catalog No. AA4680103

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 26570-48-9
    Molecular Formula CH2=COC(OCH2CH2)nOCOC=CH2
    MDL Number MFCD00081876
    Synonym PEG-diacrylate
    Chemical Name or Material Polyethylene glycol diacrylate, M.W. 8,000

    Specifications

    Physical Form Granular Solid
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H317-H335-H315
    Causes serious eye damage.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Causes skin irritation.
    Color White to off-white
    CAS 26570-48-9
    Synonym PEG-diacrylate
    Recommended Storage 2°C to 8°C
    Quantity 1g
    TSCA TSCA
    Molecular Formula CH2=COC(OCH2CH2)nOCOC=CH2
    MDL Number MFCD00081876
    Chemical Name or Material Polyethylene glycol diacrylate, M.W. 8,000

    GHS H Statement
    H318-H317-H335-H315
    Causes serious eye damage.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Causes skin irritation.

    2°C to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit