Alfa Aesar™ Polyethylene glycol methyl ether acrylate, M.W. 5,000

Manufacturer: Alfa Aesar™ 46388MD

Quantity 250mg

 View more versions of this product

Catalog No. AA46388MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 32171-39-4
    Molecular Formula CH3(OCH2CH2)nOCOC=CH2
    MDL Number MFCD00803685
    Synonym mPEG-acrylate
    Chemical Name or Material Polyethylene glycol methyl ether acrylate, M.W. 5,000

    Specifications

    Physical Form Granular Solid
    Synonym mPEG-acrylate
    TSCA TSCA
    Molecular Formula CH3(OCH2CH2)nOCOC=CH2
    MDL Number MFCD00803685
    CAS 32171-39-4
    Recommended Storage 2°C to 8°C
    Color White to off-white
    Quantity 250mg
    Chemical Name or Material Polyethylene glycol methyl ether acrylate, M.W. 5,000

    2°C to 8°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit