Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol Mono-4-nonylphenyl Ether (n=approx. 5), TCI America™

Click to view available options
Quantity:
25g
26027-38-3
MFCD00148006
26027-38-3
MFCD00148006
Color Yellow
Quantity 25g
Physical Form Clear Liquid at 20°C
Chemical Name or Material Polyethylene Glycol Mono-4-nonylphenyl Ether (n=approx. 5)

EINECSNumber : (7)-0172

RTECSNumber : RB1248636

TSCA : Yes