Alfa Aesar™ Polyethylene, high density

Manufacturer: Alfa Aesar™ 41731A1

Quantity 1kg

 View more versions of this product

Catalog No. AA41731A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 9002-88-4
    MDL Number MFCD00084423
    Merck Index 14,7567
    Chemical Name or Material Polyethylene, High density

    Specifications

    TSCA TSCA
    Density 0.95g/mL
    Physical Form Granules
    MDL Number MFCD00084423
    CAS 9002-88-4
    Merck Index 14,7567
    Quantity 1kg
    Odor Odorless
    Chemical Name or Material Polyethylene, High density

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit