Alfa Aesar™ Polyethylene oxide, M.W. 300,000

Manufacturer: Alfa Aesar™ 4223714

Quantity 25g

 View more versions of this product

Catalog No. AA4223714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 25322-68-3
    MDL Number MFCD00147979
    Chemical Name or Material Polyethylene oxide, M.W. 300,000

    Specifications

    Melting Point 65°C
    RTECS Number TQ3630000
    Physical Form Powder
    MDL Number MFCD00147979
    Odor Odorless
    Refractive Index 1.4539
    Color White to off-white
    pH 5 to 7
    Quantity 25g
    TSCA TSCA
    Density 1.21g/mL
    CAS 25322-68-3
    Chemical Name or Material Polyethylene oxide, M.W. 300,000

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit