Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Polyethylene oxide, M.W. >5,000,000

Click to view available options
Quantity:
100g
500g
25322-68-3
(-CH2CH2O-)n
MFCD00147992
25322-68-3
(-CH2CH2O-)n
MFCD00147992
Solubility Information Soluble in water.
Formula Weight >5,000,000
Odor Odorless
Melting Point ~175°C
pH 5 to 7
Quantity 500g
Chemical Name or Material Polyethylene oxide

EINECSNumber : 500-038-2

TSCA : Yes

Recommended Storage : Ambient temperatures