Alfa Aesar™ Polyethylene sheet, High Density, 1.6mm (0.063 in.) thick

Manufacturer: Alfa Aesar™ 45155HB

Quantity 300x300mm

 View more versions of this product

Catalog No. AA45155HB

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 9002-88-4
    MDL Number MFCD00084423
    Chemical Name or Material Polyethylene sheet, High Density, 1.6mm (0.063 in.) thick

    Specifications

    TSCA TSCA
    MDL Number MFCD00084423
    CAS 9002-88-4
    Odor Odorless
    Color White
    Quantity 300x300mm
    Chemical Name or Material Polyethylene sheet, High Density, 1.6mm (0.063 in.) thick

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit